Få mere at vide
Bromfenac Sodium Sesquihydrate, TRC
Artikelnummer. 30137981
Skift visning
Mængde:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137981 10 mg
17910635 250 mg
17914175 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137981 Leverandør Toronto Research Chemicals Leverandørnr. TRCB67855010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 120638-55-3
Molekylær formel 2C15H11BrNO3.2Na.3H2O
Molekylvægt (g/mol) 766.34
Synonym Benzeneacetic acid, 2-amino-3-(4-bromobenzoyl)-, sodium salt, hydrate (2:2:3), Benzeneacetic acid, 2-amino-3-(4-bromobenzoyl)-, monosodium salt, hydrate (2:3) (9CI)
IUPAC navn disodium;2-[2-amino-3-(4-bromobenzoyl)phenyl]acetate;trihydrate
SMIL O.O.O.[Na+].[Na+].Nc1c(CC(=O)[O-])cccc1C(=O)c2ccc(Br)cc2.Nc3c(CC(=O)[O-])cccc3C(=O)c4ccc(Br)cc4
Tekniske data
Kemisk navn eller materiale Bromfenac Sodium Sesquihydrate
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/2C15H12BrNO3.2Na.3H2O/c2*16-11-6-4-9(5-7-11)15(20)12-3-1-2-10(14(12)17)8-13(18)19;;;;;/h2*1-7H,8,17H2,(H,18,19);;;3*1H2/q;;2*+1;;;/p-2
Formel vægt 763.9957
Procent renhed >95%