Få mere at vide
Brofaromine, MedChemExpress
Artikelnummer. 16455658
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
Pakningsstørrelse:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16455658 10 mg 10mg
16418357 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16455658 Leverandør Medchem Express Leverandørnr. HY13339
MedChemExpress Brofaromine (CGP 11305A) is a monoamine oxidase (MAO) inhibitor with IC50 of 0.2 μM for MAO-A.
Brofaromine (CGP 11305A) is a monoamine oxidase (MAO) inhibitor with IC50 of 0.2 μM for MAO-A.
Tekniske data
Kemisk navn eller materiale Brofaromine
CAS 63638-91-5
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C14H16BrNO2
Mængde 10 mg
Synonym CGP 11305A
Opløselighedsinformation DMSO : 250 mg/mL (805.96 mM; Need ultrasonic)
SMIL BrC1=CC(OC)=CC2=C1OC(C3CCNCC3)=C2
Molekylvægt (g/mol) 310.19
Formel vægt 310.19
Procent renhed 98.55%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation