missing translation for 'onlineSavingsMsg'
Learn More

Brivudine, 98%

CAS: 69304-47-8 | C11H13BrN2O5 | 333.14 g/mol

493.00 DKK - 4155.00 DKK

Chemical Identifiers

CAS 69304-47-8
Molecular Formula C11H13BrN2O5
Molecular Weight (g/mol) 333.14
MDL Number MFCD00058585
InChI Key ODZBBRURCPAEIQ-PIXDULNESA-N
Synonym brivudine, bvdu, brivudin, helpin, e-5-2-bromovinyl-2'-deoxyuridine, bromovinyldeoxyuridine, zostex, brivudine inn, brivudinum inn-latin, brivudina inn-spanish
PubChem CID 446727
IUPAC Name 5-[(E)-2-bromoethenyl]-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(\C=C\Br)C(=O)NC1=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11387449
SDS View Documents
Thermo Scientific Alfa Aesar
H27853.MF
50 mg
493.00 DKK
50mg
Please to purchase this item. Need a web account? Register with us today!
15443797
SDS View Documents
Thermo Scientific Alfa Aesar
H27853.MD
250 mg
1375.00 DKK
250mg
Please to purchase this item. Need a web account? Register with us today!
11397449
SDS View Documents
Thermo Scientific Alfa Aesar
H27853.03
1 g
4155.00 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in water and methanol.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : YU7355000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brivudine, 98%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.