Få mere at vide
Brigatinib, MedChemExpress
Artikelnummer. 16491587
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16491587 50 mg -
16488117 5 mg -
16476006 100 mg 100mg
16435408 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16491587 Leverandør Medchem Express Leverandørnr. HY12857
MedChemExpress Brigatinib (AP-26113) is a highly potent and selective ALK inhibitor, with an IC50 of 0.6 nM.
Brigatinib (AP-26113) is a highly potent and selective ALK inhibitor, with an IC50 of 0.6 nM.
Tekniske data
Kemisk navn eller materiale Brigatinib
CAS 1197953-54-0
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C29H39ClN7O2P
Mængde 50 mg
Synonym AP-26113
Opløselighedsinformation Ethanol : 10 mg/mL (17.12 mM; Need ultrasonic and warming) ∣DMSO : 2 mg/mL (3.42 mM; Need ultrasonic)
SMIL CN1CCN(C2CCN(C3=CC=C(NC4=NC=C(Cl)C(NC5=CC=CC=C5P(C)(C)=O)=N4)C(OC)=C3)CC2)CC1
Molekylvægt (g/mol) 584.09
Formel vægt 584.09
Procent renhed 98.96%
Grad Research
Fysisk form Powder
Til brug med (applikation) Cancer-Kinase/protease