missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Borosilicate Glass Replacement Impeller

Made of borosilicate glass with two large sidearms and screw cap closures for easy sampling

Brand:  Wheaton™ 356847

 View more versions of this product

Product Code. 11591285

  • 1930.00 DKK / Each
Only null left
Estimated Shipment: 17-11-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Magna-Flex™ microcarrier spinner flasks feature a flex type, bulb shaped glass impeller which rotates from a fixed position above the liquid level around an indentation in the base of the flask. Made of borosilicate glass with two large sidearms and screw cap closures for easy sampling. 500mL and larger units have 45mm sidearms to be used as convenient vents and ports. All flasks provide a headspace ratio of 1:1 or greater to provide maximum surface interface between culture and vessel atmosphere. A removable stainless steel pin immobilises the impeller during handling. Replacement glass impeller assemblies are available for all flasks. For use with microcarrier cultures.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Borosilicate Glass
Two large sidearms and screw cap
Replacement Impeller
3000mL
Microcarrier cultures
1 Each
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Wheaton™ Borosilicate Glass Replacement Impeller > Volume Working: 3000mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.