Learn More
Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
215.00 DKK - 603.00 DKK
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | ||||||
10120181
|
Lenz Laborglasinstrumente™
01400045 |
NS 45/40 |
603.00 DKK
Pack of 5 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
10517371
|
Lenz Laborglasinstrumente™
01400034 |
NS 34/35 |
384.00 DKK
Pack of 5 |
Estimated Shipment: 15-08-2025
Log in to see stock. |
||||||
10291631
|
Lenz Laborglasinstrumente™
01400007 |
NS 7/16 |
215.00 DKK
Pack of 10 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
Description
- Acc. to DIN 12252, form C
- Available in a range of cone sizes
- Corrosion-free and resistant to thermal shock
Specifications
Borosilicate Glass | |
Hollow Stopper |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate