Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
Manufacturer:  Lenz Laborglasinstrumente™ 01400034
Additional Details : Weight : 0.10000kg
 View more versions of this product
Catalog No. 10517371
  • 384.00 DKK / Pack of 5
Only null left
Estimated Shipment: 14-07-2025
Add to cart
Description
Description
  • Acc. to DIN 12252, form C
  • Available in a range of cone sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
5/Pk.
NS 34/35
Borosilicate Glass
Hollow Stopper