Learn More
Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 04006128
 View more versions of this product
Product Code. 10393511
  • 676.00 DKK / Pack of 2
Only null left
Estimated Shipment: 09-07-2025
Description
  • With scale (approx. volume/mL)
  • With PTFE plug
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
50 mL
Borosilicate Glass
2Pack
Polyethylene
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale > Capacity: 50mL