missing translation for 'onlineSavingsMsg'
Learn More
Boric Acid, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10682251

Boric Acid, Certified AR for Analysis, Fisher Chemical™

Product Code. p-7072867 Shop All Fisher Chemical Products
500 g, 500g
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Unit Size:
1kg
2.50kg
500g
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 10682251

Brand: Fisher Chemical B/3800/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

H3BO3, CAS Number-10043-35-3, 500g, White, 61.83g/mol, 169 deg.C, 11337, Plastic powder jar, 3.8 to 4.8, Powder Solid, 2.7mbar at 20 deg.C | CAS: 10043-35-3 | BH3O3 | 61.83 g/mol

H3BO3, CAS Number-10043-35-3, 500g, White, 61.83g/mol, 169 deg.C, 11337, Plastic powder jar, 3.8 to 4.8, Powder Solid, 2.7mbar at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8 to 4.8
Color White
Melting Point 169°C
Packaging Plastic powder jar
Vapor Pressure 2.7mbar at 20°C
Quantity 500 g
Physical Form Powder Solid
Formula Weight 61.83g/mol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot Number2538592 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeB/3800/60,B/3800/70,B/3800/53,B/3800/62
Lot Number2535090 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeB/3800/60,B/3800/70,B/3800/53,B/3800/62
compliance-icons
Product Identifier
  • Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : boric acid
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156

    Expires2025-06-30

    Days

    2

    Hours

    20

    Minutes

    44

    Seconds

    10
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boric Acid, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.