Learn More
Thermo Scientific™ Bonded AVCS Pre-slit Caps
Significantly reduce the error rate of analysis that results from evaporation of solvent in vials which are not sealed correctly.
Brand: Thermo Scientific™ C5000-72G
Additional Details : Weight : 0.03000kg
Description
Significantly reduce the error rate of analysis that results from evaporation of solvent in vials which are not sealed correctly.
- Re-analysis costs are reduced, sample data integrity increases, and sample throughput increases.
- AVCS closures seal as well as crimp top vials.
- AVCS design allows for consistent sealing, eliminating user-to-user variance.
Specifications
Green | |
Yes | |
100/Pk. |
9 mm | |
PTFE/Silicone |