Learn More
Thermo Scientific™ Bonded AVCS Pre-slit Caps
Significantly reduce the error rate of analysis that results from evaporation of solvent in vials which are not sealed correctly.
289.68 DKK - 339.59 DKK
Specifications
Diameter (Metric) Outer | 9 mm |
---|---|
Pre-slit | Yes |
Septum | PTFE/Silicone |
Quantity | 100/Pk. |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
15328036
|
Thermo Scientific™
C5000-72Y |
Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15318036
|
Thermo Scientific™
C5000-72R |
Red |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15398026
|
Thermo Scientific™
C5000-72G |
Green |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15378026
|
Thermo Scientific™
C5000-72B |
Blue |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Significantly reduce the error rate of analysis that results from evaporation of solvent in vials which are not sealed correctly.
- Re-analysis costs are reduced, sample data integrity increases, and sample throughput increases.
- AVCS closures seal as well as crimp top vials.
- AVCS design allows for consistent sealing, eliminating user-to-user variance.
Specifications
9 mm | |
Yes | |
PTFE/Silicone | |
100/Pk. |