Få mere at vide
Boc-D-4-Iodophenylalanine, TRC
Artikelnummer. 30008207
Skift visning
Mængde:
500 mg
1 g
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30008207 500 mg -
30008969 2.5 g 2.5g
30003901 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008207 Leverandør Toronto Research Chemicals Leverandørnr. TRCB658948500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 176199-35-2
Molekylær formel C14 H18 I N O4
Molekylvægt (g/mol) 391.202
IUPAC navn (2R)-3-(4-iodophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMIL CC(C)(C)OC(=O)N[C@H](Cc1ccc(I)cc1)C(=O)O
Tekniske data
Kemisk navn eller materiale Boc-D-4-Iodophenylalanine
InChI formel InChI=1S/C14H18INO4/c1-14(2,3)20-13(19)16-11(12(17)18)8-9-4-6-10(15)7-5-9/h4-7,11H,8H2,1-3H3,(H,16,19)(H,17,18)/t11-/m1/s1
Mængde 500 mg
Formel vægt 391.028