missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BMPR1B (ALK6), GST Tag Recombinant Protein
Human BMPR1B (ALK6), GST Tag Recombinant Protein
3660.00 DKK - 55700.00 DKK
Specifications
Protein Length | aa 149-end |
---|---|
Species | Human |
Recombinant | Recombinant |
Protein Tag | GST Tag |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15498893
|
Invitrogen™
PV6236 |
100μg |
12470.00 DKK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15488893
|
Invitrogen™
PV6235 |
10μg |
3660.00 DKK
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15408903
|
Invitrogen™
PV6237 |
1mg |
55700.00 DKK
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
BMPR1B is a member of the transmembrane serine/threonine kinase that is the member of the bone morphogenetic protein (BMP) receptor. BMPR1B is mainly involved in the endochondral bone formation and embryogenesis. BMPR1B expressed in normal and cancerous prostate tissues and used in the endocrine therapy that given to the prostate cancer patients. BMPR1B receptor trafficking also play a significant role in FOP pathogenesis and used in human T-cell differentiation.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
aa 149-end | |
Human | |
Recombinant | |
GST Tag |