missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BMPR1B (ALK6), GST Tag Recombinant Protein

Human BMPR1B (ALK6), GST Tag Recombinant Protein

Brand:  Invitrogen™ PV6237

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15408903

  • 55700.00 DKK / 1mg
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

BMPR1B is a member of the transmembrane serine/threonine kinase that is the member of the bone morphogenetic protein (BMP) receptor. BMPR1B is mainly involved in the endochondral bone formation and embryogenesis. BMPR1B expressed in normal and cancerous prostate tissues and used in the endocrine therapy that given to the prostate cancer patients. BMPR1B receptor trafficking also play a significant role in FOP pathogenesis and used in human T-cell differentiation.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
66.7kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Human
GST Tag
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
1mg
aa 149-end
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers