missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
BLZF1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16354-1-AP-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The BLZF1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human BLZF1. This antibody recognizes human, mouse antigen. The BLZF1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Tekniske data
| BLZF1 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8548 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 350 μg/mL | |
| GOLGIN-45; JEM-1; JEM-1s; JEM1; MGC22497 | |
| Fusion Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 45 kDa | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion