Learn More
BRAND™ Blaubrand™ ETERNA™ Class A Volumetric Flasks, with Certificate
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. Calibrated to contain a precise amount of liquid at a specific temperature. Includes polypropylene stopper and DE-M marking. Delivered with individual conformity certificate.
403.00 DKK - 449.00 DKK
Specifications
Calibration | Calibrated To Contain |
---|---|
Closure Material | Polypropylene |
Color | Clear |
Material | Borosilicate Glass |
Graduated | Yes |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
15825532
|
BRAND™
936838 |
5 mL |
403.00 DKK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
15845532
|
BRAND™
936843 |
10 mL |
404.00 DKK
Each |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
15835532
|
BRAND™
936842 |
25 mL |
406.00 DKK
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
||||||
15855532
|
BRAND™
936845 |
20 mL |
406.00 DKK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||||
15865532
|
BRAND™
936849 |
100 mL |
427.00 DKK
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
15875532
|
BRAND™
936851 |
250 mL |
449.00 DKK
Each |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
Description
- Class A Volumetric Flask
- Constructed of Borosilicate Glass
- Complies with DIN EN ISO 1042 standards
- Calibrated to contain (TC, In)
- Narrow neck and wide neck options available in a variety of sizes
- Includes polypropylene stopper and DE-M marking Delivered with individual conformity certificate
Specifications
Calibrated To Contain | |
Clear | |
Yes | |
Stopper |
Polypropylene | |
Borosilicate Glass | |
A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ Blaubrand™ ETERNA™ Class A Volumetric Flasks, with Certificate