Få mere at vide
Bisphenol TMC, TRC
Artikelnummer. 30134710
Skift visning
Mængde:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134710 25 g
30011327 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134710 Leverandør Toronto Research Chemicals Leverandørnr. TRCB51957025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 129188-99-4
Molekylær formel C21 H26 O2
Molekylvægt (g/mol) 310.43
Synonym 4,4'-(3,3,5-Trimethylcyclohexylidene)bis[phenol], 1,1-Bis(4-hydroxyphenyl)-3,3,5-trimethylcyclohexane, 3,3,5-Trimethyl-1,1-bis(4-hydroxyphenyl)cyclohexane, 4,4'-(3,3,5-Trimethylcyclohexylidene)diphenol, BISP-TMC, BPTMC, Bisphenol TMC
IUPAC navn 4-[1-(4-hydroxyphenyl)-3,3,5-trimethylcyclohexyl]phenol
SMIL CC1CC(C)(C)CC(C1)(c2ccc(O)cc2)c3ccc(O)cc3
Tekniske data
Kemisk navn eller materiale Bisphenol TMC
Renhedsgrad noter HPLC
Mængde 25 g
InChI formel InChI=1S/C21H26O2/c1-15-12-20(2,3)14-21(13-15,16-4-8-18(22)9-5-16)17-6-10-19(23)11-7-17/h4-11,15,22-23H,12-14H2,1-3H3
Formel vægt 310.1933
Procent renhed >95%