Få mere at vide
Bisphenol P, TRC
Artikelnummer. 17962345
Skift visning
Mængde:
250 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17962345 25 g
17953285 5 g
17963685 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17962345 Leverandør Toronto Research Chemicals Leverandørnr. TRCB51947525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2167-51-3
Molekylær formel C24 H26 O2
Molekylvægt (g/mol) 346.46
Synonym Phenol, 4,4'-(p-phenylenediisopropylidene)di- (7CI,8CI), 4,4'-[1,4-Phenylenebis(1-methylethylidene)]bis[phenol], 1,4-Bis(1-(4-hydroxyphenyl)-1-methylethyl)benzene, 1,4-Bis(4-hydroxycumyl)benzene, 1,4-Bis(p-hydroxycumyl)benzene, 1,4-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene, 1,4-Bis[α-methyl-α-(4-hydroxyphenyl)ethyl]benzene, 4,4'-(p-Phenylenediisopropylidene)bisphenol, 4,4'-(p-Phenylenediisopropylidene)diphenol, B 1563, Bisphenol P, α,α'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene, α,α'-Bis(4-hydroxyphenyl)-p-diisopropylbenzene
IUPAC navn 4-[2-[4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl]propan-2-yl]phenol
SMIL CC(C)(c1ccc(O)cc1)c2ccc(cc2)C(C)(C)c3ccc(O)cc3
Tekniske data
Kemisk navn eller materiale Bisphenol P
Mængde 25 g
InChI formel InChI=1S/C24H26O2/c1-23(2,19-9-13-21(25)14-10-19)17-5-7-18(8-6-17)24(3,4)20-11-15-22(26)16-12-20/h5-16,25-26H,1-4H3
Formel vægt 346.1933