missing translation for 'onlineSavingsMsg'
Få mere at vide

Bisphenol F Glycidyl 2,3-Dihydroxypropyl Ether, TRC

Artikelnummer. 30012360
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012360 100 mg
30007786 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30012360 Leverandør Toronto Research Chemicals Leverandørnr. TRCB519585100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303733-72-4
Molekylær formel C19H22O5
Molekylvægt (g/mol) 330.37
Synonym 3-[4-[[4-(2-Oxiranylmethoxy)phenyl]methyl]phenoxy]-1,2-propanediol
IUPAC navn 3-[4-[[4-(oxiran-2-ylmethoxy)phenyl]methyl]phenoxy]propane-1,2-diol
SMIL OCC(O)COc1ccc(Cc2ccc(OCC3CO3)cc2)cc1

Specifications

Kemisk navn eller materiale Bisphenol F Glycidyl 2,3-Dihydroxypropyl Ether
Mængde 100 mg
InChI formel InChI=1S/C19H22O5/c20-10-16(21)11-22-17-5-1-14(2-6-17)9-15-3-7-18(8-4-15)23-12-19-13-24-19/h1-8,16,19-21H,9-13H2
Formel vægt 330.1467
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.