Få mere at vide
Bisphenol F Glycidyl 2,3-Dihydroxypropyl Ether, TRC
Artikelnummer. 30007786
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007786 10 mg
30012360 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30007786 Leverandør Toronto Research Chemicals Leverandørnr. TRCB51958510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 303733-72-4
Molekylær formel C19H22O5
Molekylvægt (g/mol) 330.37
Synonym 3-[4-[[4-(2-Oxiranylmethoxy)phenyl]methyl]phenoxy]-1,2-propanediol
IUPAC navn 3-[4-[[4-(oxiran-2-ylmethoxy)phenyl]methyl]phenoxy]propane-1,2-diol
SMIL OCC(O)COc1ccc(Cc2ccc(OCC3CO3)cc2)cc1
Tekniske data
Kemisk navn eller materiale Bisphenol F Glycidyl 2,3-Dihydroxypropyl Ether
Mængde 10 mg
InChI formel InChI=1S/C19H22O5/c20-10-16(21)11-22-17-5-1-14(2-6-17)9-15-3-7-18(8-4-15)23-12-19-13-24-19/h1-8,16,19-21H,9-13H2
Formel vægt 330.1467