Få mere at vide
Bis(aminoethoxy) Amlodipine, TRC
Artikelnummer. 30011751
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011751 10 mg
30006685 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011751 Leverandør Toronto Research Chemicals Leverandørnr. TRCB40210010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 721958-74-3
Molekylær formel C22 H30 Cl N3 O6
Molekylvægt (g/mol) 467.94
Synonym 3,5-Pyridinedicarboxylic acid, 2,6-bis[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-, 3-ethyl 5-methyl ester, 3,5-Pyridinedicarboxylic acid, 2,6-bis[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-, ethyl methyl ester (9CI)
IUPAC navn 5-O-ethyl 3-O-methyl 2,6-bis(2-aminoethoxymethyl)-4-(2-chlorophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMIL CCOC(=O)C1=C(COCCN)NC(=C(C1c2ccccc2Cl)C(=O)OC)COCCN
Tekniske data
Kemisk navn eller materiale Bis(aminoethoxy) Amlodipine
Mængde 10 mg
InChI formel InChI=1S/C22H30ClN3O6/c1-3-32-22(28)20-17(13-31-11-9-25)26-16(12-30-10-8-24)19(21(27)29-2)18(20)14-6-4-5-7-15(14)23/h4-7,18,26H,3,8-13,24-25H2,1-2H3
Formel vægt 467.1823