Learn More
Bis(4-morpholinyl)methane, 98% Product Code.: 15408017
Bis(4-morpholinyl)methane, 98%
Product Code. 15408017
25 g, 25g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15408017
Brand: Thermo Scientific Alfa Aesar H51133.14
Request Bulk or Custom Format
CAS: 5625-90-1 | C9H18N2O2 | 186.255 g/mol
It is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as intermediate for pharmaceutical.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5625-90-1
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD00023369
InChI Key MIFZZKZNMWTHJK-UHFFFAOYSA-N
Synonym dimorpholinomethane, n,n'-dimorpholinomethane, 4,4-methylenedimorpholine, 4,4'-methylenedimorpholine, bismorpholino methane, n,n'-methylenebismorpholine, morpholine, 4,4'-methylenebis, unii-7o79dzw79z, bis morpholino-methan german, bis 4-morpholinyl methane
PubChem CID 21839
IUPAC Name 4-(morpholin-4-ylmethyl)morpholine
SMILES C1COCCN1CN2CCOCC2
Specifications
Boiling Point 122°C to 124°C (12 mmHg)
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 186.25
Percent Purity 98%
Chemical Name or Material Bis(4-morpholinyl)methane
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberP03H010 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH51133.06,H51133.14
Lot NumberM23D016 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH51133.06,H51133.14
Lot NumberR22D054 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH51133.06,H51133.14
Lot NumberN22H091 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH51133.06,H51133.14
Lot NumberX19H006 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH51133.06,H51133.14
RUO – Research Use Only
Product Content Correction
Product Title
Bis(4-morpholinyl)methane, 98% >