missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) phthalate 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30311226
Change view
Click to view available options
Mængde:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30311226 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30311226 Supplier Dr. Ehrenstorfer Supplier No. DREGA09011139ME

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Phthalic acid, bis-2-ethylhexyl ester
CAS 117-81-7
Molekylær formel C24 H38 O4
Mængde 1 mL
Synonym Diethylhexylphthalate, Bis-(2-ethylhexyl) Phthalate, Plastic additive 01, DEHP
InChI formel InChI=1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3
SMIL CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC
IUPAC navn bis(2-ethylhexyl) benzene-1,2-dicarboxylate
Molekylvægt (g/mol) 390.56
Formel vægt 390.277 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.