missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Betulinic acid

CAS: 472-15-1 | C30H48O3 | 456.71 g/mol

939.00 DKK - 2985.00 DKK

Chemical Identifiers

CAS 472-15-1
Molecular Formula C30H48O3
Molecular Weight (g/mol) 456.71
InChI Key QGJZLNKBHJESQX-FZFNOLFKSA-N
PubChem CID 64971
ChEBI CHEBI:3087
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15881012
View Documents
Thermo Scientific Chemicals
462872500
250 mg
939.00 DKK
250mg
Please to purchase this item. Need a web account? Register with us today!
15871012
View Documents
Thermo Scientific Chemicals
462870010
1 g
2985.00 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

472-15-1
456.71
64971
C30H48O3
QGJZLNKBHJESQX-FZFNOLFKSA-N
CHEBI:3087

Specifications

472-15-1
QGJZLNKBHJESQX-FZFNOLFKSA-N
64971
Betulinic acid
C30H48O3
456.71
CHEBI:3087
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Betulinic acid

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.