Få mere at vide
Bethanechol chloride, 98%, Thermo Scientific™
A cholinergic receptor agonist
Brand: Thermo Scientific Alfa Aesar J65919.06
| Mængde | 5g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| C7H17ClN2O2 | |
| MFCD00055224 | |
| bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone | |
| CHEBI:3085 | |
| CC(C[N+](C)(C)C)OC(=O)N.[Cl-] |
Tekniske data
| Bethanechol chloride | |
| C7H17ClN2O2 | |
| 5g | |
| bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone | |
| CC(C[N+](C)(C)C)OC(=O)N.[Cl-] | |
| 196.675 | |
| CHEBI:3085 | |
| 98% |
| 590-63-6 | |
| MFCD00055224 | |
| 14,1185 | |
| XXRMYXBSBOVVBH-UHFFFAOYSA-N | |
| 2-carbamoyloxypropyl(trimethyl)azanium;chloride | |
| 11548 | |
| 196.68 |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.