Learn More
Thermo Scientific™ BetaSil™ Phenyl HPLC Columns, 5μm Particle Size
Thermo ScientificTM BetaSilTM Phenyl HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
Brand: Thermo Scientific™ 70605-102130
Description
Thermo ScientificTM BetaSilTM Phenyl HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
- Thermo Scientific chemistry on 100Ă… silica
Ordering Information:
Other column dimensions including preparative scale are available. Please call Customer Service for more information.
Specifications
Analytical Column | |
LC/MS | |
5 ÎĽm | |
100 Ă… | |
Phenyl (Ph) | |
Silica | |
Reversed Phase | |
Reversed Phase |
2.1 mm | |
100 mm | |
2 to 9 | |
L11 | |
Beta | |
11% | |
Yes |