Få mere at vide
Betamethasone 21-O-Ethyl Carbonate, TRC
Artikelnummer. 30138155
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138155 100 mg
30136658 250 mg
30004007 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138155 Leverandør Toronto Research Chemicals Leverandørnr. TRCB327120100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 52619-05-3
Molekylær formel C25 H33 F O7
Molekylvægt (g/mol) 464.52
Synonym Pregna-1,4-diene-3,20-dione, 21-[(ethoxycarbonyl)oxy]-9-fluoro-11,17-dihydroxy-16-methyl-, (11beta,16beta)-, (11beta,16beta)-21-[(Ethoxycarbonyl)oxy]-9-fluoro-11,17-dihydroxy-16-methylpregna-1,4-diene-3,20-dione, Betamethasone EP Impurity D
IUPAC navn ethyl [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] carbonate
SMIL CCOC(=O)OCC(=O)[C@@]1(O)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
Tekniske data
Kemisk navn eller materiale Betamethasone 21-(ethyl carbonate)
Mængde 100 mg
InChI formel InChI=1S/C25H33FO7/c1-5-32-21(30)33-13-20(29)25(31)14(2)10-18-17-7-6-15-11-16(27)8-9-22(15,3)24(17,26)19(28)12-23(18,25)4/h8-9,11,14,17-19,28,31H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1
Formel vægt 464.221