missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ 8 HPLC Columns, 5ÎĽm Particle Size

Thermo ScientificTM BetaBasicTM 8 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.

Brand:  Thermo Scientific™ 71405-052130

6074.34 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10417064

  • 6395.00 DKK / Each
Only null left
In Stock
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo ScientificTM BetaBasicTM 8 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.

  • Pore size suitable for small molecules, peptides and protein digests
  • Superb pH stability

Ordering Information:

Other column dimensions are also available. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Analytical Column
LC/MS, APCI
5 ÎĽm
150 Ă…
L7
Silica
Reversed Phase
Reversed Phase
2.1 mm
50 mm
2 to 9
200 m²/g
C8
7%
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ BetaBasic™ 8 HPLC Columns, 5ÎĽm Particle Size > Particle Size: 5ÎĽm; 50 x 2.1mm I.D.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.