missing translation for 'onlineSavingsMsg'
Få mere at vide

Beta-Methylcholine Chloride, TRC

Artikelnummer. 30133909
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133909 10 g
30137028 5 g
30010126 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30133909 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30585510G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2382-43-6
Molekylær formel C6H16ClNO
Molekylvægt (g/mol) 153.65
Synonym beta-Hydroxypropyltrimethylammonium Chloride, (2-Hydroxypropyl)trimethylammonium Chloride, 2-Hydroxy-N,N,N-trimethyl-1-propanaminium Chloride;
IUPAC navn 2-hydroxypropyl(trimethyl)azanium;chloride
SMIL [Cl-].CC(O)C[N+](C)(C)C

Specifications

Kemisk navn eller materiale beta-Methylcholine Chloride
Mængde 10 g
InChI formel InChI=1S/C6H16NO.ClH/c1-6(8)5-7(2,3)4;/h6,8H,5H2,1-4H3;1H/q+1;/p-1
Formel vægt 153.092
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.