missing translation for 'onlineSavingsMsg'
Få mere at vide

Benzyl Violet (Technical Grade), TRC

Artikelnummer. 17904335
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17904335 500 mg
17952755 250 mg
17931645 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17904335 Leverandør Toronto Research Chemicals Leverandørnr. TRCB316553500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1694-09-3
Molekylær formel C39 H40 N3 O6 S2 . Na
Molekylvægt (g/mol) 733.87
Synonym Acid Violet 5B (6CI), Benzenemethanaminium, N-[4-[[4-(dimethylamino)phenyl][4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-, hydroxide, inner salt, sodium salt, Benzenemethanaminium, N-[4-[[4-(dimethylamino)phenyl][4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-, inner salt, sodium salt (9CI), C.I. Acid Violet 49 (7CI), C.I. Acid Violet 49, sodium salt (8CI), 11386 Violet, A.F. Violet No. 1, Acid Fast Violet 5BN, Acid Milling Violet 4BS, Acid Violet, Acid Violet 23184, Acid Violet 49, Acid Violet 4BNP, Acid Violet 5BN, Acid Violet 6B, Acid Violet S, Acilan Violet S 4BN, Aizen Acid Violet 5BH, Atlantic Acid Violet 4BNS, Benzyl Violet, Benzyl Violet 3B, Benzyl Violet 4B, Borunil Violet A-6B, C.I. 42640, C.I. Food Violet 2, Cogilor Violet 411.12, Coomassie Violet, D And C Violet No. 1, Dinacid Violet 4B, Dycosacid Violet 5B, Everacid Violet 4BR, FD and C Violet 1, FD and C Violet No. 1, FD&C Violet No. 1, Fast Acid Violet 5BN, Food Violet 2, Hidacid Wool Violet 5B, Ichoacid Violet M, Indacid Violet S 4BN, Kenanthrol Violet 4BNS, Kiton Violet 4BNS, Libacid Violet L4BS, Lowalan Violet 49, Monacid Violet 4BS, Old Japan Purple 1, Old Japan Violet 1, Orbacid Violet S 4B, Orient Water Violet 1, Pacid Violet 4BS, Pergacid Violet 2B, Ratna Acid Violet 5BND, Ravi Acid Violet 4B, Serva Violet 49, Tertracid Brilliant Violet 6B, Triacid Violet 4BN, Violet 6B, Violet No. 1, Water Violet 1, Wool Violet 4BN, Wool Violet 5BN
IUPAC navn sodium;3-[[4-[[4-(dimethylamino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate
SMIL [Na+].CCN(Cc1cccc(c1)S(=O)(=O)[O-])c2ccc(cc2)C(=C3C=CC(=[N+](CC)Cc4cccc(c4)S(=O)(=O)[O-])C=C3)c5ccc(cc5)N(C)C

Tekniske data

Kemisk navn eller materiale Acid Violet 49
Mængde 500 mg
InChI formel InChI=1S/C39H41N3O6S2.Na/c1-5-41(27-29-9-7-11-37(25-29)49(43,44)45)35-21-15-32(16-22-35)39(31-13-19-34(20-14-31)40(3)4)33-17-23-36(24-18-33)42(6-2)28-30-10-8-12-38(26-30)50(46,47)48;/h7-26H,5-6,27-28H2,1-4H3,(H-,43,44,45,46,47,48);/q;+1/p-1
Formel vægt 733.2256
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.