missing translation for 'onlineSavingsMsg'
Learn More
Benzethonium chloride, 97% Product Code.: 11403197

Benzethonium chloride, 97%

Product Code. p-7023079
100 g, 100g
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 11403197

Brand: Thermo Scientific Alfa Aesar A16142.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

CAS: 121-54-0 | C27H42ClNO2 | 448.09 g/mol

Benzethonium chloride is used as a surface disinfectant and a preservative in food processing and pharmaceutical industries. It is involved in the determination of sodium dodecyl sulfate using sulphan blue as indicator from the mixture containing other ingredients like sodium chloride and sodium sulfate. It is an effective cationic detergent due to its positively charged hydrophilic part of quaternary ammonium ion. It is an active ingredient in medicaments, deodorants and mouthwashes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzethonium chloride is used as a surface disinfectant and a preservative in food processing and pharmaceutical industries. It is involved in the determination of sodium dodecyl sulfate using sulphan blue as indicator from the mixture containing other ingredients like sodium chloride and sodium sulfate. It is an effective cationic detergent due to its positively charged hydrophilic part of quaternary ammonium ion. It is an active ingredient in medicaments, deodorants and mouthwashes.

Solubility
Soluble in water.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-54-0
Molecular Formula C27H42ClNO2
MDL Number MFCD00011742
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
Synonym benzethonium chloride, quatrachlor, hyamine, phemeride, phemerol chloride, benzethoniumchloride, phemithyn, disilyn, kylacol, solamin
PubChem CID 8478
ChEBI CHEBI:31264

Specifications

Melting Point 157°C to 165°C
Odor Odorless
Quantity 100 g
UN Number UN1759
Beilstein 3898548
Sensitivity Hygroscopic
Merck Index 14,1074
Solubility Information Soluble in water.
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]
IUPAC Name benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
Molecular Weight (g/mol) 448.088
Formula Weight 448.1
Percent Purity 97%
Chemical Name or Material Benzethonium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberP25L041 Certificate TypeCertificate of Analysis Date27/03/2025 Product CodeA16142.0E,A16142.36,A16142.22
Lot NumberX02K026 Certificate TypeCertificate of Analysis Date10/10/2024 Product CodeA16142.36,A16142.22
Lot NumberZ19J044 Certificate TypeCertificate of Analysis Date16/01/2024 Product CodeA16142.36,A16142.22
Lot NumberZ06J037 Certificate TypeCertificate of Analysis Date04/01/2024 Product CodeA16142.36,A16142.22
Lot NumberW19J019 Certificate TypeCertificate of Analysis Date04/10/2023 Product CodeA16142.0E,A16142.36,A16142.22
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzethonium chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Benzethonium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzethonium chloride, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.