Learn More
Cytiva Benchkote sheet for ÄKTA systems
Benchkote sheets for ÄKTA systems will protect the top buffer trays from buffer spillages and salt deposits.
143.00 DKK - 316.00 DKK
Specifications
Material | Absorbent Paper, Laminated Polyethylene |
---|---|
Description | For use with ÄKTA go |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
16497645
|
Cytiva
2300-10094 |
25/Pk. |
316.00 DKK
Pack of 50 |
|
||||||
16477645
|
Cytiva
2300-10092 |
10/Pk. |
143.00 DKK
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
16487645
|
Cytiva
2300-10093 |
50/Pk. |
204.00 DKK
Pack of 25 |
Estimated Shipment: 07-11-2025
Log in to see stock. |
||||||
Description
Why Benchkote sheet for lab ÄKTA systems?
- Cover the top buffer tray with Benchkote sheet and protect from buffer spillages and salt deposits.
- Simplify daily and weekly preventive maintenance of the top buffer tray of your ÄKTA systems
- Smooth, absorbent Whatman™ paper quickly absorbs spills.
- Laminated polyethylene layer prevents flow-through to the working surface.
- Offered in three convenient pack sizes: 10/pk, 25/pk and 50/pk sheets.
- Convenient and easy to use.
- Benchkote sheets are manufactured of absorbent, impermeable material designed to protect lab surfaces against hazardous spills.
- The high-quality, smooth, and absorbent Whatman™ paper, quickly absorbs liquid spills, and laminated polyethylene layer prevents flow-through to the working surface.
Specifications
Absorbent Paper, Laminated Polyethylene | |
For use with ÄKTA go |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Cytiva Benchkote sheet for ÄKTA systems