missing translation for 'onlineSavingsMsg'
Få mere at vide

BDE 28, TRC

Artikelnummer. 17949339
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
5 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17949339 25 mg -
17927139 5 mg -
17952055 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17949339 Leverandør Toronto Research Chemicals Leverandørnr. TRCB11921025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 41318-75-6
Molekylær formel C12 H7 Br3 O
Molekylvægt (g/mol) 406.9
Synonym Benzene, 2,4-dibromo-1-(4-bromophenoxy)-, Ether, p-bromophenyl 2,4-dibromophenyl (6CI), 2,4-Dibromo-1-(4-bromophenoxy)benzene, 2,4,4'-Tribromodiphenyl ether, BDE 28, PBDE 28
IUPAC navn 2,4-dibromo-1-(4-bromophenoxy)benzene
SMIL Brc1ccc(Oc2ccc(Br)cc2Br)cc1

Tekniske data

Kemisk navn eller materiale PBDE 28 (2,4,4'-Tribromodiphenyl Ether)
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C12H7Br3O/c13-8-1-4-10(5-2-8)16-12-6-3-9(14)7-11(12)15/h1-7H
Formel vægt 403.8047
Procent renhed >95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.