Learn More
Brady™ BBP™ Pre-Printed Right-To-Know Chemical Labels with Color Bar with text area
Allows printer to produce five-color chemical labels in a single print pass
2375.00 DKK - 2932.40 DKK
Specifications
Certifications/Compliance | RoHS 2002/95/EC |
---|---|
Adhesive Type | Acrylic, Permanent, Pressure-sensitive |
Color | Red, Yellow, Blue, White |
Format | Label |
Material | Vinyl |
Product Code | Brand | Size (English) | Size (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (English) | Size (Metric) | Quantity | Price | Quantity & Availability | |||
11747264
|
Brady™
142043 |
2.25 H x 4 in. W | 57.2 H x 101.6 mm W | 240/Pk. |
2375.00 DKK
1 box |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||
11757334
|
Brady™
142040 |
2.25 in. H x 2.25 in. W | 57.2 mm H x 57.2 mm W | 365/Pk. |
2375.00 DKK
1 box |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||
11727174
|
Brady™
142039 |
4 in. H x 6.25 in. W | 101.6 mm H x 158.8 mm W | 170/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Indoor/outdoor-grade vinyl film with permanent adhesive
- Material withstands exposure to moisture, chemicals and handling
- Available in multiple sizes and formats
- Service Temp.: -40° to +82°C (-40° to +180°F)
- Application Temp.: 18° to 82°C (0° to 180°F)
- Outdoor Durability: 8 to 10 years
Specifications
RoHS 2002/95/EC | |
Red, Yellow, Blue, White | |
Vinyl | |
Pre-cut blank sign headers allow the printer to create multicolor signs fast - in a single print pass, conforms to rough, irregular and “difficult to adhere to” surfaces | |
60 to 50 Gardner Units | |
-40 to 180°F | |
0.004 in. |
Acrylic, Permanent, Pressure-sensitive | |
Label | |
Color Bar With Text Area | |
BBP30, BBP31, BBP33, BBP35, and BBP37 printers | |
8 to 10 Years | |
-40°C to +82°C |