Learn More
RETSCH BB 200 Model Jaw Crushers
Brand: RETSCH 20.058.1002
Specifications
Jaw Crusher | |
900 x 450 x 1160 mm | |
Suitable for integration in automatic installations | |
IP54 | |
Stainless Steel | |
1-phase/3-phase |
CE Certified | |
Voltages on request | |
1160 mm | |
900 mm | |
BB 200 Version for Integration | |
450 mm |