Få mere at vide
Barasertib-HQPA, MedChemExpress
Artikelnummer. 16496027
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16496027 5 mg -
16484556 100 mg 100mg
16473308 10 mg -
16459846 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16496027 Leverandør Medchem Express Leverandørnr. HY10126
MedChemExpress Barasertib-HQPA (AZD2811) is a highly selective Aurora B inhibitor with an IC50 of 0.37 nM in a cell-free assay. Barasertib-HQPA (AZD2811) induces growth arrest and apoptosis in cancer cells.
Barasertib-HQPA (AZD2811) is a highly selective Aurora B inhibitor with an IC50 of 0.37 nM in a cell-free assay. Barasertib-HQPA (AZD2811) induces growth arrest and apoptosis in cancer cells.
Tekniske data
Kemisk navn eller materiale Barasertib-HQPA
CAS 722544-51-6
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C26H30FN7O3
Mængde 5 mg
Synonym AZD2811 INH-34 AZD1152-HQPA
Opløselighedsinformation DMSO : ≥ 22 mg/mL (43.34 mM)
SMIL O=C(CC1=NNC(NC2=C3C=CC(OCCCN(CCO)CC)=CC3=NC=N2)=C1)NC4=CC=CC(F)=C4
Molekylvægt (g/mol) 507.56
Formel vægt 507.56
Procent renhed 98.57%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease