Få mere at vide
BAM7, MedChemExpress
Artikelnummer. 16496458
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16496458 10 mg
16432567 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16496458 Leverandør Medchem Express Leverandørnr. HY15341
MedChemExpress BAM7 is a direct and selective activator of proapoptotic BAX with an IC50 of 3.3 μM.
BAM7 is a direct and selective activator of proapoptotic BAX with an IC50 of 3.3 μM.
Tekniske data
Kemisk navn eller materiale BAM7
CAS 331244-89-4
Farve Red
Renhedsgrad noter Research
Molekylær formel C21H19N5O2S
Mængde 10 mg
Opløselighedsinformation DMSO : 5 mg/mL (12.33 mM; Need ultrasonic)
SMIL CCOC1=CC=CC=C1N/N=C2C(C)=NN(C3=NC(C4=CC=CC=C4)=CS3)C\2=O
Molekylvægt (g/mol) 405.47
Formel vægt 405.47
Procent renhed 99.18%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death