Learn More
Vitlab™ Büchner Funnels
Filter out impurities with Buchner funnel. Vitlab™ Büchner funnels are exceptional for filtration and removing impurities. The upper and lower part of the funnel is detachable to facilitate cleaning.
113.00 DKK - 158.00 DKK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10778101
|
Vitlab™
80437 |
40 mL |
113.00 DKK
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
10604271
|
Vitlab™
80438 |
70 mL |
121.00 DKK
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
10429582
|
Vitlab™
80440 |
280 mL |
158.00 DKK
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Description
- Polypropylene construction
- Upper and lower parts are detachable
Specifications
Polypropylene |