Learn More
Vitlab™ Büchner Funnels
Filter out impurities with Buchner funnel. Vitlab™ Büchner funnels are exceptional for filtration and removing impurities. The upper and lower part of the funnel is detachable to facilitate cleaning.
Brand: Vitlab™ 80438
Additional Details : Weight : 0.01000kg
Description
- Polypropylene construction >
- Upper and lower parts are detachable
Specifications
70 mL | |
55mm | |
113mm |
Polypropylene | |
1.1mm |