Learn More
Azlon™ Class B PMP Volumetric Flask
Manufactured from transparent PMP provides clear easy to read calibration line
Brand:  Azlon™ FDF112N
 View more versions of this product
Product Code. 10554345
  • 193.00 DKK / Each
Only null left
Estimated Shipment: 16-10-2025
Description
    • Calibrated to ISO 1042, class B tolerances
    • Supplied with polypropylene stopper
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transparent
PMP
1
Autoclavable
Stopper
25 mL
115 mm (with stopper)
±0.08 mL
B
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Azlon™ Class B PMP Volumetric Flask > Capacity: 25mL