Få mere at vide
AZD1390, MedChemExpress
Artikelnummer. 16481129
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
10 mg
25 mg
50 mg
Pakningsstørrelse:
25mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16481129 1 mg -
16424308 10 mg -
16403036 25 mg 25mg
16400637 50 mg -
16417027 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16481129 Leverandør Medchem Express Leverandørnr. HY109566
MedChemExpress AZD1390 is a potent, highly selective, orally bioavailable, brain-penetrant ATM inhibitor with an IC50 of 0.78 nM in cell.
AZD1390 is a potent, highly selective, orally bioavailable, brain-penetrant ATM inhibitor with an IC50 of 0.78 nM in cell.
Tekniske data
Kemisk navn eller materiale AZD1390
CAS 2089288-03-7
Farve White
Renhedsgrad noter Research
Molekylær formel C27H32FN5O2
Mængde 1 mg
Opløselighedsinformation DMSO : 25 mg/mL (52.35 mM; Need ultrasonic and warming)
SMIL O=C(N1C(C)C)N(C)C2=C1C3=CC(C4=CC=C(OCCCN5CCCCC5)N=C4)=C(F)C=C3N=C2
Molekylvægt (g/mol) 477.57
Formel vægt 477.57
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease