missing translation for 'onlineSavingsMsg'
Få mere at vide

AZ304, MedChemExpress

Artikelnummer. 16435656
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16435656 100 mg
16431127 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16435656 Leverandør Medchem Express Leverandørnr. HY117273

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress AZ304 is an ATP-competitive dual BRAF kinase inhibitor, potently inhibits wild type BRAF, V600E mutant BRAF and wild type CRAF, with IC50s of 79 nM, 38 nM and 68 nM, respectively. AZ304 also has significant effect on other kinases, such as p38 (IC50, 6 nM), CSF1R (IC50, 35 nM). Anti-tumor activity.

AZ304 is an ATP-competitive dual BRAF kinase inhibitor, potently inhibits wild type BRAF, V600E mutant BRAF and wild type CRAF, with IC50s of 79 nM, 38 nM and 68 nM, respectively. AZ304 also has significant effect on other kinases, such as p38 (IC50, 6 nM), CSF1R (IC50, 35 nM). Anti-tumor activity.

Tekniske data

Kemisk navn eller materiale AZ304
CAS 942507-42-8
Farve White
Renhedsgrad noter Research
Molekylær formel C27H25N5O2
Mængde 100 mg
Opløselighedsinformation DMSO : 125 mg/mL (276.84 mM; Need ultrasonic)
SMIL O=C(NC1=CC=C(C)C(NC2=C3C=CC(OC)=CC3=NC=N2)=C1)C4=CC=CC(C(C)(C#N)C)=C4
Molekylvægt (g/mol) 451.52
Formel vægt 451.52
Procent renhed 99.96%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.