missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Clear Stacked Format Automation Tips for Hamilton Microlab™ Star Line and Nimbus™
For optimum performance with various automated liquid handler systems
8920.00 DKK
Specifications
Filtered | Non-Filtered |
---|---|
Color | Clear |
Autoclavable | Not Autoclavable |
Clean Claims | DNase, RNase and Pyrogen free |
For Use With (Application) | Non-pyrogenic, used for stringent specifications under strict process controls |
Description
Axygen offers a comprehensive line of tips for Hamilton™ Microlab™ automation systems.
The tips are:
- Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
- RNase-/DNase-free and non-pyrogenic
- Available in non-sterile, sterile, non-filtered and filtered
- Available in 96-well hanging tip rack and nested tip rack and individual deck-seating rack format
- Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
Specifications
Non-Filtered | |
Not Autoclavable | |
Non-pyrogenic, used for stringent specifications under strict process controls | |
Automation, Round | |
Polypropylene |
Clear | |
DNase, RNase and Pyrogen free | |
Stack Rack | |
4 Racks/Stack, 30 Stacks/Cs. |