missing translation for 'onlineSavingsMsg'
Få mere at vide

AX-024, MedChemExpress

Artikelnummer. 16482956
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16482956 25 mg -
16470477 50 mg -
16406827 5 mg -
16405096 100 mg -
16404128 10 mg 10mg
16400126 2 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16482956 Leverandør Medchem Express Leverandørnr. HY107390

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress AX-024 is an orally available, first-in-class inhibitor of the TCR-Nck interaction that selectively inhibits TCR-triggered T cell activation with an IC50 ∼1 nM. AX-024 modulates cell signaling by targeting SH3 domains. AX-024 has low-acute toxicity and high potency and selectivity, and strongly inhibit the production of IL-6, TNF-α, IFN-γ, IL-10 and IL-17A.

AX-024 is an orally available, first-in-class inhibitor of the TCR-Nck interaction that selectively inhibits TCR-triggered T cell activation with an IC50 ∼1 nM. AX-024 modulates cell signaling by targeting SH3 domains. AX-024 has low-acute toxicity and high potency and selectivity, and strongly inhibit the production of IL-6, TNF-α, IFN-γ, IL-10 and IL-17A.

Tekniske data

Kemisk navn eller materiale AX-024
CAS 1370544-73-2
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C21H22FNO2
Mængde 25 mg
Opløselighedsinformation Ethanol : 100 mg/mL (294.64 mM; Need ultrasonic) ∣DMSO : 35 mg/mL (103.12 mM; Need ultrasonic and warming)
SMIL COC1=CC=C(OC2)C(C(C3=CC=C(F)C=C3)=C2CN4CCCC4)=C1
Molekylvægt (g/mol) 339.4
Formel vægt 339.4
Procent renhed 98.0%
Grad Research
Fysisk form Powder
Til brug med (applikation) COVID-19-immunoregulation
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.