Få mere at vide
Bollé Safety Attack Clear Safety Goggle
Shop alle Bollé Safety produkter
:
Frame Color: Clear
Beskrivelse
- Soft compression face seal
- Fitted with a single carbon-plus treated polycarbonate lens
- Fog and scratch resistant on both sides
Tekniske data
Tekniske data
| Linsebelægning | Anti-fog, Scratch-resistant |
| Linsefarve | Clear |
| nr. pr. pakke | 1 Ea. |
| Vægt (metrisk) | 91 g |