Få mere at vide
Atropine-N-oxide Hydrochloride, TRC
Artikelnummer. 30136186
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136186 100 mg
30134019 500 mg
30009688 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136186 Leverandør Toronto Research Chemicals Leverandørnr. TRCA793860100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4574-60-1
Molekylær formel C17 H23 N O4 . Cl H
Molekylvægt (g/mol) 341.83
Synonym Benzeneacetic acid, α-(hydroxymethyl)-, (3-endo)-8-methyl-8-oxido-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (1:1), 1αH,5αH-Tropan-3α-ol, (±)-tropate (ester), 8-oxide, hydrochloride (8CI), Benzeneacetic acid, α-(hydroxymethyl)-, (3-endo)-8-methyl-8-oxido-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (9CI), Benzeneacetic acid, α-(hydroxymethyl)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester endo-(±)-, N-oxide, hydrochloride, Atropine N-oxide hydrochloride, Atropine aminoxide hydrochloride, Atropine oxide hydrochloride, Genatropine hydrochloride, Tropinox, Xtro
IUPAC navn [(1R,5S)-8-methyl-8-oxido-8-azoniabicyclo[3.2.1]octan-3-yl] (2R)-3-hydroxy-2-phenylpropanoate;hydrochloride
SMIL Cl.C[N+]1([O-])[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@@H](CO)c3ccccc3
Tekniske data
Kemisk navn eller materiale Atropine N-Oxide Hydrochloride
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C17H23NO4.ClH/c1-18(21)13-7-8-14(18)10-15(9-13)22-17(20)16(11-19)12-5-3-2-4-6-12;/h2-6,13-16,19H,7-11H2,1H3;1H/t13-,14+,15+,16-,18?;/m0./s1
Formel vægt 341.1394
Procent renhed >95%