Få mere at vide
Atorvastatin Cyclic Sodium Salt (Isopropyl) Impurity, TRC
Artikelnummer. 30010253
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010253 5 mg
30010024 1 mg
30007386 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010253 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7918255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1316291-19-6
Molekylær formel C33 H34 F N2 O7 . Na
Molekylvægt (g/mol) 612.62
Synonym 7-(4-Fluorophenyl)hexahydro-beta,7-dihydroxy-1b-(1-methylethyl)-7a-phenyl-1a-[(phenylamino)carbonyl]-3H-oxireno[3,4]pyrrolo[2,1-b][1,3]oxazine-3-butanoic Acid Sodium Salt (1:1)
IUPAC navn sodium;4-[7-(4-fluorophenyl)-7-hydroxy-7a-phenyl-1a-(phenylcarbamoyl)-1b-propan-2-yl-4,5-dihydro-3H-oxireno[1,2]pyrrolo[4,5-b][1,3]oxazin-3-yl]-3-hydroxybutanoate
SMIL [Na+].CC(C)C12OC(CC(O)CC(=O)[O-])CCN1C(O)(c3ccc(F)cc3)C4(OC24C(=O)Nc5ccccc5)c6ccccc6
Tekniske data
Kemisk navn eller materiale Atorvastatin Cyclic Sodium Salt (Isopropyl) Impurity
Analyt- eller komponentnavne Atorvastatin Cyclic Sodium Salt (Isopropyl) Impurity
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C33H35FN2O7.Na/c1-21(2)33-31(29(40)35-25-11-7-4-8-12-25)30(43-31,22-9-5-3-6-10-22)32(41,23-13-15-24(34)16-14-23)36(33)18-17-27(42-33)19-26(37)20-28(38)39;/h3-16,21,26-27,37,41H,17-20H2,1-2H3,(H,35,40)(H,38,39);/q;+1/p-1
Løsningstype Neat
Industry Type Forensics
Mængde 5 mg
Formel vægt 612.2248
Procent renhed >95%