Få mere at vide
AST 487, MedChemExpress
Artikelnummer. 16488867
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
Pakningsstørrelse:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16488867 5 mg 5mg
16472407 50 mg -
16436288 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16488867 Leverandør Medchem Express Leverandørnr. HY15002
MedChemExpress AST 487 is a RET kinase inhibitor with IC50 of 880 nM, inhibits RET autophosphorylation and activation of downstream effectors, also inhibits Flt-3 with IC50 of 520 nM.
AST 487 is a RET kinase inhibitor with IC50 of 880 nM, inhibits RET autophosphorylation and activation of downstream effectors, also inhibits Flt-3 with IC50 of 520 nM.
Tekniske data
Kemisk navn eller materiale AST 487
CAS 630124-46-8
Farve White
Renhedsgrad noter Research
Molekylær formel C26H30F3N7O2
Mængde 5 mg
Synonym NVP-AST 487
Opløselighedsinformation DMSO : ≥ 100 mg/mL (188.84 mM)
SMIL O=C(NC1=CC=C(OC2=NC=NC(NC)=C2)C=C1)NC3=CC=C(CN4CCN(CC)CC4)C(C(F)(F)F)=C3
Molekylvægt (g/mol) 529.56
Formel vægt 529.56
Procent renhed 99.2%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease