Få mere at vide
IBA Solutions For Life Science™ ApU-dinucleotide IBA
Artikelnummer. 12889366 Shop alle IBA Lifesciences produkter
Skift visning
Mængde:
10 mg
Pakningsstørrelse:
10mg
Artikelnummer. Mængde Pakningsstørrelse
12889366 10 mg 10mg
1 options
Artikelnummer. 12889366 Leverandør IBA Solutions For Life Science™ Leverandørnr. 50710147
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Dinucleotides are used by RNA polymerases as initiators for RNA synthesis. Therefore, they are very useful for transcription analysis.