missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
APD668, MedChemExpress
Description
APD668 is a potent, selective and orally active agonist of G-protein coupled receptor GPR119, with EC50s of 2.7 nM and 33 nM for hGPR119 and rGPR119, respectively. APD668 shows no significant inhibition of any of the five major CYP isoforms with the exception of CYP2C9 (Ki=0.1 μM). APD668 can be used for the research of steatohepatitis and diabetes.
Specifications
Specifications
| Kemisk navn eller materiale | APD668 |
| CAS | 832714-46-2 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C21H24FN5O5S |
| Mængde | 5 mg |
| Opløselighedsinformation | DMSO : 33.33 mg/mL (69.80 mM; Need ultrasonic) |
| SMIL | O=C(OC(C)C)N(CC1)CCC1OC2=C3C(N(C4=CC=C(S(=O)(C)=O)C=C4F)N=C3)=NC=N2 |
| Molekylvægt (g/mol) | 477.51 |
| Formel vægt | 477.51 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?